Gene Transcripts

Gene Symbol
Pds5a
Entrez Gene ID
305343
Gene Name
PDS5 cohesin associated factor A
Gene Aliases
RGD1307094, Scc-112
Chromosome Location
Chr. 14: 44185445 - 44282308 on Build Rnor_6.0
UniGene ID
Rn.224580
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083624.1 NP_001077093.1 14 1597
XM_008770153.1 XP_008768375.1 14 1631
XM_008770154.1 XP_008768376.1 13 1544
GenBank mRNA EF460313.1 - 14 1597

View Full Product Details