Gene Transcripts

Gene Symbol
Mfrp
Entrez Gene ID
315597
Gene Name
membrane frizzled-related protein
Gene Aliases
Chromosome Location
Chr. 8: 48437814 - 48443421 on Build Rnor_6.0
UniGene ID
Rn.20302
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108137.2 NP_001101607.1 11 1502
XM_006242933.2 XP_006242995.1 11 1929

View Full Product Details