Gene Transcripts

Gene Symbol
Lsm14a
Entrez Gene ID
361554
Gene Name
LSM14A, SCD6 homolog A (S. cerevisiae)
Gene Aliases
RGD1305695
Chromosome Location
Chr. 1: 90200866 - 90246292 on Build Rnor_6.0
UniGene ID
Rn.2370
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127552.1 NP_001121024.1 8 1131
XM_006228856.2 XP_006228918.1 8 1179
XM_006228857.2 XP_006228919.1 8 1179
XM_006228858.2 XP_006228920.1 7 1002
GenBank mRNA BC166537.1 - 8 1131

View Full Product Details