Gene Transcripts

Gene Symbol
Ralgapb
Entrez Gene ID
362257
Gene Name
Ral GTPase activating protein, beta subunit (non-catalytic)
Gene Aliases
RGD1306861, p170
Chromosome Location
Chr. 3: 154910295 - 154983021 on Build Rnor_6.0
UniGene ID
Rn.28469
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271210.1 NP_001258139.1 17 2195
XM_006235415.2 XP_006235477.1 17 2604
XM_006235416.2 XP_006235478.1 16 2595
XM_006235417.2 XP_006235479.1 17 2592
XM_006235418.2 XP_006235480.1 16 2583
XM_006235420.2 XP_006235482.1 15 2559
XM_006235421.2 XP_006235483.1 15 2547
XM_008762357.1 XP_008760579.1 16 2619
XM_008762358.1 XP_008760580.1 15 2583

View Full Product Details