Gene Transcripts

Gene Symbol
Rtel1
Entrez Gene ID
362288
Gene Name
regulator of telomere elongation helicase 1
Gene Aliases
RGD1306721
Chromosome Location
Chr. 3: 176818024 - 176856531 on Build Rnor_6.0
UniGene ID
Rn.98315
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191857.1 NP_001178786.1 29 2940
XM_006235772.2 XP_006235834.1 29 2996
XM_006235773.2 XP_006235835.1 29 3405
XM_006235774.2 XP_006235836.1 29 2996
XM_006235775.2 XP_006235837.1 29 2996
XM_006235776.2 XP_006235838.1 29 2996
XM_006235778.2 XP_006235840.1 22 2535
GenBank mRNA BC086436.1 - 2 98
BC161910.1 - 5 490

View Full Product Details