Gene Transcripts

Gene Symbol
Seli
Entrez Gene ID
362713
Gene Name
selenoprotein I
Gene Aliases
Ept1, RGD1560938
Chromosome Location
Chr. 6: 27473748 - 27512287 on Build Rnor_6.0
UniGene ID
Rn.64052
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134754.1 NP_001128226.2 1, 2 144
GenBank mRNA BC168235.1 - 1, 2 144

View Full Product Details