Gene Transcripts

Gene Symbol
Ranbp9
Entrez Gene ID
364686
Gene Name
RAN binding protein 9
Gene Aliases
RGD1561940, RanBPM
Chromosome Location
Chr. 17: 24020874 - 24103771 on Build Rnor_6.0
UniGene ID
Rn.155466
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771586.1 XP_008769808.1 15 2243
GenBank mRNA FQ218715.1 - 2 191

View Full Product Details