Gene Transcripts

Gene Symbol
Ankdd1b
Entrez Gene ID
100359662
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
Chromosome Location
Chr. 2: 27030295 - 27296860 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760686.1 XP_008758908.1 10 1078

View Full Product Details