Gene Transcripts

Gene Symbol
Prkcg
Entrez Gene ID
24681
Gene Name
protein kinase C, gamma
Gene Aliases
PKC, PKCI, Prkc, Prkcc, RATPKCI
Chromosome Location
Chr. 1: 64407098 - 64433703 on Build Rnor_6.0
UniGene ID
Rn.9747
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012628.1 NP_036760.1 8 1068
XM_006228014.2 XP_006228076.1 8 1136
GenBank mRNA BC089226.1 - 8 1102
M13707.1 - 8 1068
X07287.1 - 8 991

View Full Product Details