Gene Transcripts

Gene Symbol
Lipogenin
Entrez Gene ID
252964
Gene Name
Lipogenin
Gene Aliases
Chromosome Location
Chr. 4: 56033448 - 56034703 on Build Rnor_6.0
UniGene ID
Rn.212748
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145790.1 NP_665733.1 1 997
GenBank mRNA AY057076.1 - 1 997

View Full Product Details