Gene Transcripts

Gene Symbol
Trim80
Entrez Gene ID
287824
Gene Name
tripartite motif protein 80
Gene Aliases
Trim47, Trim47l
Chromosome Location
Chr. 10: 103984642 - 104004060 on Build Rnor_6.0
UniGene ID
Rn.198897
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768412.1 XP_008766634.1 6 2982
GenBank mRNA BC091694.1 - 5 956

View Full Product Details