Gene Transcripts

Gene Symbol
Chmp6
Entrez Gene ID
287873
Gene Name
charged multivesicular body protein 6
Gene Aliases
RGD1565325
Chromosome Location
Chr. 10: 109073678 - 109079348 on Build Rnor_6.0
UniGene ID
Rn.7707
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105856.1 NP_001099326.1 2, 3 222
GenBank mRNA BC070963.1 - 14

View Full Product Details