Gene Transcripts

Gene Symbol
Vom2r69
Entrez Gene ID
289433
Gene Name
vomeronasal 2 receptor, 69
Gene Aliases
RGD1564750
Chromosome Location
Chr. 14: 1007495 - 1020982 on Build Rnor_6.0
UniGene ID
Rn.217643
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099468.1 NP_001092938.1 3 997

View Full Product Details