Gene Transcripts

Gene Symbol
Klk9
Entrez Gene ID
292851
Gene Name
kallikrein related-peptidase 9
Gene Aliases
Chromosome Location
Chr. 1: 99729704 - 99733996 on Build Rnor_6.0
UniGene ID
Rn.218609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106253.1 NP_001099723.1 3 441

View Full Product Details