Gene Transcripts

Gene Symbol
Map7
Entrez Gene ID
293016
Gene Name
microtubule-associated protein 7
Gene Aliases
Mtap7
Chromosome Location
Chr. 1: 15620665 - 15748203 on Build Rnor_6.0
UniGene ID
Rn.23807
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106270.2 NP_001099740.2 4 498
NM_001198638.1 NP_001185567.1 4 274
GenBank mRNA CB767890.1 - 4 274

View Full Product Details