Gene Transcripts

Gene Symbol
Zfyve26
Entrez Gene ID
314265
Gene Name
zinc finger, FYVE domain containing 26
Gene Aliases
Chromosome Location
Chr. 6: 102409235 - 102472962 on Build Rnor_6.0
UniGene ID
Rn.16048
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108038.1 NP_001101508.1 37 7129
XM_008764781.1 XP_008763003.1 37 7165

View Full Product Details