Gene Transcripts

Gene Symbol
Ttc7b
Entrez Gene ID
362768
Gene Name
tetratricopeptide repeat domain 7B
Gene Aliases
Chromosome Location
Chr. 6: 124316444 - 124528058 on Build Rnor_6.0
UniGene ID
Rn.64438
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108719.2 NP_001102189.1 3 297
XM_008764816.1 XP_008763038.1 9 1007
GenBank mRNA BC168996.1 - 3 297
FQ210625.1 - 5 425

View Full Product Details