Gene Transcripts

Gene Symbol
RT1-CE10
Entrez Gene ID
414792
Gene Name
RT1 class I, locus CE10
Gene Aliases
Chromosome Location
Chr. 20: 154072 - 157679 on Build Rnor_6.0
UniGene ID
Rn.219012
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008833.1 NP_001008833.1 2 273
XM_006255847.2 XP_006255909.1 2 278
GenBank mRNA BC083874.1 - 2 276
BC098952.1 - 2 276 7366301

View Full Product Details