Gene Transcripts

Gene Symbol
Emr4
Entrez Gene ID
450235
Gene Name
EGF-like module containing, mucin-like, hormone receptor-like sequence 4
Gene Aliases
Emr4p
Chromosome Location
Chr. 9: 8056052 - 8199945 on Build Rnor_6.0
UniGene ID
Rn.162355
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007558.1 NP_001007559.1 841
XM_008766767.1 XP_008764989.1 829
GenBank mRNA AY686633.1 - 841

View Full Product Details