Gene Transcripts

Gene Symbol
Vom1r23
Entrez Gene ID
494288
Gene Name
vomeronasal 1 receptor 23
Gene Aliases
V1rf5
Chromosome Location
Chr. 1: 61685997 - 61686944 on Build Rnor_6.0
UniGene ID
Rn.129445
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008941.1 NP_001008941.1 1 817
GenBank mRNA AY510359.1 - 1 817

View Full Product Details