Gene Transcripts

Gene Symbol
Itga2b
Entrez Gene ID
685269
Gene Name
integrin, alpha 2B
Gene Aliases
Chromosome Location
Chr. 10: 90397960 - 90416544 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001063315.5 XP_001063315.1 29 4447

View Full Product Details