Gene Transcripts

Gene Symbol
Tprg1l
Entrez Gene ID
687090
Gene Name
tumor protein p63 regulated 1-like
Gene Aliases
mover
Chromosome Location
Chr. 5: 171456299 - 171459488 on Build Rnor_6.0
UniGene ID
Rn.202609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110318.2 NP_001103788.1 3 633
GenBank mRNA BC160865.1 - 3 482
EU220430.1 - 3 633

View Full Product Details