Gene Transcripts

Gene Symbol
Nufip2
Entrez Gene ID
681371
Gene Name
nuclear fragile X mental retardation protein interacting protein 2
Gene Aliases
Chromosome Location
Chr. 10: 65175344 - 65200215 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750867.3 XP_003750915.1 2 1929

Gene Symbol
RGD1564405
Entrez Gene ID
500386
Gene Name
similar to mKIAA1321 protein
Gene Aliases
Chromosome Location
Chr. 5: 3675824 - 3678446 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749882.3 XP_003749930.1 2 1760

View Full Product Details