Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NADK2
Entrez Gene ID
704285
Gene Name
NAD kinase 2, mitochondrial
Gene Aliases
C6H5orf33, NADKD1
Chromosome Location
Chr.6: 35989627 - 36035149 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001092624.4 XP_001092624.2 - 431 131
XM_015139873.2 XP_014995359.1 - 432 131
XM_028849386.1 XP_028705219.1 - 116 131
XM_028849387.1 XP_028705220.1 - 228 131
XM_028849388.1 XP_028705221.1 - 116 131

View Full Product Details