Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PLA2G12A
- Entrez Gene ID
-
697726
- Gene Name
- phospholipase A2 group XIIA
- Gene Aliases
-
EGK_16009
- Chromosome Location
-
Chr.5: 108069530 - 108081207 on Build Mmul_10
- Gene Symbol
- DCN
- Entrez Gene ID
-
709933
- Gene Name
- decorin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 90914795 - 90952424 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV046313.1 |
- |
- |
702 |
72 |
View Full Product Details