Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBK
Entrez Gene ID
713242
Gene Name
PDZ binding kinase
Gene Aliases
-
Chromosome Location
Chr.8: 28077588 - 28100995 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001109768.4 XP_001109768.1 - 735 74
XM_002805290.3 XP_002805336.1 - 822 74
GenBank mRNA JU476400.1 - - 769 74

View Full Product Details