Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FZD3
Entrez Gene ID
713839
Gene Name
frizzled class receptor 3
Gene Aliases
EGK_18811, Frizzled-3
Chromosome Location
Chr.8: 28756486 - 28828121 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261141.1 NP_001248070.1 - 2139 66
GenBank mRNA JU336417.1 - - 2139 66

View Full Product Details