Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5DC3
Entrez Gene ID
699285
Gene Name
5'-nucleotidase domain containing 3
Gene Aliases
-
Chromosome Location
Chr.11: 103480601 - 103544631 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015152619.2 XP_015008105.2 - 804 65
GenBank mRNA JU327270.1 - - 619 65
JV653445.1 - - 734 65

View Full Product Details