Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX3
Entrez Gene ID
700730
Gene Name
peroxisomal biogenesis factor 3
Gene Aliases
EGK_15288
Chromosome Location
Chr.4: 47229225 - 47286046 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257927.1 NP_001244856.1 - 1129 105
GenBank mRNA JU321855.1 - - 1129 105
JU477567.1 - - 1085 105
JV043457.1 - - 1129 105
JV527836.1 - - 1145 105

View Full Product Details