Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOSR1
Entrez Gene ID
574268
Gene Name
golgi SNAP receptor complex member 1
Gene Aliases
-
Chromosome Location
Chr.16: 26147989 - 26200848 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257945.2 NP_001244874.1 - 368 87
XM_015118837.2 XP_014974323.1 - 455 87
XM_015118838.2 XP_014974324.1 - 455 87
XM_015118839.2 XP_014974325.1 - 396 87
XM_028835453.1 XP_028691286.1 - 455 87
XM_028835454.1 XP_028691287.1 - 455 87
XM_028835455.1 XP_028691288.1 - 449 87
GenBank mRNA AY369815.1 - - 138 87
JU337209.1 - - 369 87
JU337210.1 - - 375 87

View Full Product Details