Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P3H1
Entrez Gene ID
697917
Gene Name
prolyl 3-hydroxylase 1
Gene Aliases
LEPRE1
Chromosome Location
Chr.1: 181790508 - 181810377 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258160.1 NP_001245089.1 - 671 86
GenBank mRNA JU471357.1 - - 671 86
JV638254.1 - - 692 86

View Full Product Details