Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC2A7
Entrez Gene ID
722147
Gene Name
solute carrier family 2 member 7
Gene Aliases
-
Chromosome Location
Chr.1: 215468365 - 215492726 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001118335.4 XP_001118335.2 - 57 60

View Full Product Details