Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EOMES
Entrez Gene ID
704711
Gene Name
eomesodermin
Gene Aliases
-
Chromosome Location
Chr.2: 20018480 - 20025222 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015132320.2 XP_014987806.2 - 1294 83
XM_015132321.2 XP_014987807.2 - 1294 83
GenBank mRNA JU494435.1 - - 123 83

View Full Product Details