Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC38A7
Entrez Gene ID
707671
Gene Name
solute carrier family 38 member 7
Gene Aliases
EGK_12847
Chromosome Location
Chr.20: 45916959 - 45940731 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260850.1 NP_001247779.1 - 1047 61
XM_028840506.1 XP_028696339.1 - 1318 61
XM_028840507.1 XP_028696340.1 - 1358 61
XM_028840509.1 XP_028696342.1 - 1653 61
XM_028840510.1 XP_028696343.1 - 1276 61
XM_028840511.1 XP_028696344.1 - 1636 61
XM_028840512.1 XP_028696345.1 - 1735 61
GenBank mRNA JU324243.1 - - 1224 61
JV045532.1 - - 1047 61
JV655227.1 - - 260 61

View Full Product Details