Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALX1
Entrez Gene ID
700027
Gene Name
ALX homeobox 1
Gene Aliases
CART1
Chromosome Location
Chr.11: 84895182 - 84918144 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015152375.2 XP_015007861.1 - 375 63

View Full Product Details