Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCHF7
Entrez Gene ID
703010
Gene Name
membrane associated ring-CH-type finger 7
Gene Aliases
EGK_04466, MARCH7
Chromosome Location
Chr.12: 46769263 - 46824317 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015110218.2 XP_014965704.1 - 2074 100
XM_015110219.2 XP_014965705.1 - 2011 100
XM_015110221.2 XP_014965707.1 - 1927 100
XM_028830742.1 XP_028686575.1 - 2074 100
XM_028830744.1 XP_028686577.1 - 2082 100
XM_028830745.1 XP_028686578.1 - 2099 100
XM_028830746.1 XP_028686579.1 - 2289 100
XM_028830747.1 XP_028686580.1 - 1906 100
XM_028830748.1 XP_028686581.1 - 1914 100
GenBank mRNA JU335959.1 - - 1904 100
JU476839.1 - - 1910 100
JV047959.1 - - 1901 100
JV591046.1 - - 1903 100

View Full Product Details