Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARID3C
Entrez Gene ID
700584
Gene Name
AT-rich interaction domain 3C
Gene Aliases
-
Chromosome Location
Chr.15: 49575855 - 49582855 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001096190.3 XP_001096190.2 - 341 84
GenBank mRNA JV571719.1 - - 210 84
JV606325.1 - - 318 84

View Full Product Details