Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOD2
Entrez Gene ID
695542
Gene Name
nucleotide binding oligomerization domain containing 2
Gene Aliases
-
Chromosome Location
Chr.20: 37792336 - 37831438 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015126107.2 XP_014981593.2 - 2476 65

View Full Product Details