Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNDIG1
Entrez Gene ID
704737
Gene Name
synapse differentiation inducing 1
Gene Aliases
EGK_02738, TMEM90B
Chromosome Location
Chr.10: 41921447 - 42127966 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194092.2 NP_001181021.1 - 737 72
XM_015149614.2 XP_015005100.1 - 1240 72
XM_015149615.2 XP_015005101.1 - 1756 72
XM_015149617.2 XP_015005103.1 - 832 72
XM_015149618.2 XP_015005104.1 - 1039 72
GenBank mRNA JU336456.1 - - 800 72
JV482424.1 - - 336 72
JV594230.1 - - 845 72
JV629203.1 - - 872 72

View Full Product Details