Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NLGN4X
Entrez Gene ID
106995148
Gene Name
neuroligin 4 X-linked
Gene Aliases
NLGN4
Chromosome Location
Chr.X: 5459345 - 5801293 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015126897.2 XP_014982383.1 - 2493 94
XM_015126898.2 XP_014982384.1 - 2857 94
XM_015126899.2 XP_014982385.1 - 2329 94
XM_028842352.1 XP_028698185.1 - 2260 94
XM_028842353.1 XP_028698186.1 - 2768 94
XM_028842354.1 XP_028698187.1 - 2299 94
XM_028842355.1 XP_028698188.1 - 1480 94
GenBank mRNA AF462606.1 - - 54 94

Gene Symbol
NLGN4Y
Entrez Gene ID
100270698
Gene Name
neuroligin 4, Y-linked
Gene Aliases
-
Chromosome Location
Chr.Y: 4141220 - 4465753 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145058.1 NP_001138530.1 - 2035 94
XM_015128588.2 XP_014984074.1 - 2078 94
XM_015128589.2 XP_014984075.1 - 2110 94
XM_015128590.2 XP_014984076.1 - 2229 94
XM_015128591.2 XP_014984077.1 - 2313 94
XM_028844192.1 XP_028700025.1 - 1550 94
XM_028844561.1 XP_028700394.1 - 1529 94
GenBank mRNA FJ527017.1 - - 2035 94
JU322425.1 - - 1937 94
JU322426.1 - - 1937 94

View Full Product Details