Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMP
Entrez Gene ID
619186
Gene Name
cathelicidin antimicrobial peptide
Gene Aliases
CAP18
Chromosome Location
Chr.2: 104256040 - 104258022 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033509.2 NP_001028681.1 - 313 63
GenBank mRNA AF181954.1 - - 316 63
JU494369.1 - - 157 63

View Full Product Details