Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ADGRE2
Entrez Gene ID
654420
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
EMR2
Chromosome Location
Chr.19: 14368627 - 14412439 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038662.1 NP_001033751.1 - 1419 66
XM_028838330.1 XP_028694163.1 - 1573 66
XM_028838331.1 XP_028694164.1 - 1573 66
XM_028838333.1 XP_028694166.1 - 1429 66
GenBank mRNA DQ227271.1 - - 1419 66
JU485338.1 - - 126 66

View Full Product Details