Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNNT2
Entrez Gene ID
709882
Gene Name
troponin T2, cardiac type
Gene Aliases
-
Chromosome Location
Chr.1: 58779791 - 58798498 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001247991.2 NP_001234920.2 - 579 78
XM_015118840.2 XP_014974326.2 - 653 78
XM_015118854.2 XP_014974340.2 - 550 78
XM_028843815.1 XP_028699648.1 - 656 78
XM_028843816.1 XP_028699649.1 - 647 78
XM_028843824.1 XP_028699657.1 - 644 78
XM_028843826.1 XP_028699659.1 - 608 78
XM_028843829.1 XP_028699662.1 - 653 78
XM_028843833.1 XP_028699666.1 - 657 78
XM_028843834.1 XP_028699667.1 - 657 78
XM_028843837.1 XP_028699670.1 - 625 78
XM_028843840.1 XP_028699673.1 - 561 78
XM_028843841.1 XP_028699674.1 - 622 78
XM_028843846.1 XP_028699679.1 - 574 78
XM_028843850.1 XP_028699683.1 - 623 78
XM_028843852.1 XP_028699685.1 - 637 78
XM_028843854.1 XP_028699687.1 - 605 78
XM_028843857.1 XP_028699690.1 - 627 78
XM_028843858.1 XP_028699691.1 - 634 78
XM_028843861.1 XP_028699694.1 - 610 78
XM_028843865.1 XP_028699698.1 - 601 78
XM_028843866.1 XP_028699699.1 - 607 78
XM_028843869.1 XP_028699702.1 - 598 78
XM_028843872.1 XP_028699705.1 - 612 78
XM_028843874.1 XP_028699707.1 - 583 78
XM_028843876.1 XP_028699709.1 - 513 78
XM_028843883.1 XP_028699716.1 - 580 78
XM_028843886.1 XP_028699719.1 - 548 78
GenBank mRNA JV556346.1 - - 449 78
JV627386.1 - - 547 78

View Full Product Details