Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA7
Entrez Gene ID
709911
Gene Name
golgin A7
Gene Aliases
EGK_18880
Chromosome Location
Chr.8: 42079358 - 42099287 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194104.1 NP_001181033.1 - 438 131
XM_015145251.2 XP_015000737.1 - 463 131
XM_015145252.2 XP_015000738.1 - 493 131
XM_015145253.2 XP_015000739.1 - 451 131
XM_015145254.2 XP_015000740.1 - 433 131
GenBank mRNA JV470468.1 - - 328 131
JV618451.1 - - 277 131

View Full Product Details