Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTAP
Entrez Gene ID
709020
Gene Name
methylthioadenosine phosphorylase
Gene Aliases
MTAPase
Chromosome Location
Chr.15: 61817872 - 61870482 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261737.1 NP_001248666.1 - 482 72
GenBank mRNA JU334602.1 - - 478 72
JU475672.1 - - 465 72
JV047118.1 - - 482 72

View Full Product Details