Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERINC5
Entrez Gene ID
710413
Gene Name
serine incorporator 5
Gene Aliases
-
Chromosome Location
Chr.6: 76950489 - 77071723 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015140335.2 XP_014995821.1 - 417 100
XM_028849546.1 XP_028705379.1 - 369 100
XM_028849547.1 XP_028705380.1 - 351 100
GenBank mRNA JU333830.1 - - 335 100
JU475038.1 - - 332 100
JV046662.1 - - 351 100
JV046663.1 - - 351 100
JV647492.1 - - 310 100

View Full Product Details