Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL2RA
Entrez Gene ID
574300
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
IL-2RA
Chromosome Location
Chr.9: 7192508 - 7242421 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032917.2 NP_001028089.1 - 580 69
GenBank mRNA AY693777.1 - - 580 69
JU472971.1 - - 786 69

View Full Product Details