Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD37
Entrez Gene ID
718718
Gene Name
CD37 molecule
Gene Aliases
-
Chromosome Location
Chr.19: 49280875 - 49286166 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001114865.4 XP_001114865.2 - 317 93
XM_028839715.1 XP_028695548.1 - 319 93
GenBank mRNA JU323266.1 - - 250 93
JU470714.1 - - 261 93

View Full Product Details