Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORD
Entrez Gene ID
712784
Gene Name
sorbitol dehydrogenase
Gene Aliases
EGK_17451
Chromosome Location
Chr.7: 21451560 - 21495438 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260778.1 NP_001247707.1 - 621 151
GenBank mRNA JU323287.1 - - 694 151
JU476912.1 - - 621 151

View Full Product Details