Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF11
Entrez Gene ID
715564
Gene Name
fibroblast growth factor 11
Gene Aliases
-
Chromosome Location
Chr.16: 7205080 - 7212164 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001108934.4 XP_001108934.1 - 2155 66

View Full Product Details